Product Details

SNP ID
rs1662059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99339221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACTTACTTATATGACAGGCAG[A/C]GGTGTCAGTGATTCCATGTTATAAT
Phenotype
MIM: 103730
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADH1C PubMed Links

Gene Details

Gene
ADH1C
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000669.4 Intron NP_000660.1
XM_011531588.2 Intron XP_011529890.1
XM_011531589.1 Intron XP_011529891.1

View Full Product Details