Product Details
- SNP ID
-
rs2172876
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128963238 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATAAACTGAAGTTCAAAGTAATA[C/G]GGTATGTGGCAAGATTCAAACCAAC
- Phenotype
-
MIM: 610032
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TNPO3
PubMed Links
Gene Details
- Gene
- TNPO3
- Gene Name
- transportin 3
View Full Product Details