Product Details
- SNP ID
-
rs34632345
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128963361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATACGATCCTGCCCTTATACTGTAA[A/C]CAGCACTGAATGGAATAATGCAAAA
- Phenotype
-
MIM: 610032
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TNPO3
PubMed Links
Gene Details
- Gene
- TNPO3
- Gene Name
- transportin 3
View Full Product Details