Product Details

SNP ID
rs4963307
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:61686718 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGGTGAGACAGGATGCTGCCTCG[A/G]ATTCATCCTGAATGGGCCCCTCGTC
Phenotype
MIM: 614015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAGLA PubMed Links

Gene Details

Gene
DAGLA
Gene Name
diacylglycerol lipase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006133.2 Intron NP_006124.1
XM_017018238.1 Intron XP_016873727.1
XM_017018239.1 Intron XP_016873728.1
XM_017018240.1 Intron XP_016873729.1

View Full Product Details