Product Details
- SNP ID
-
rs1045868
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
57
- Location
-
Chr.1:212994848 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- AAGAGAATTTAGAGCTCACTTGTCA[C/G]GAAAATATTTTTCATTATTAAAAAA
- Phenotype
-
MIM: 610471
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANGEL2
PubMed Links
Gene Details
- Gene
- ANGEL2
- Gene Name
- angel homolog 2
- Gene
- VASH2
- Gene Name
- vasohibin 2
There are no transcripts associated with this gene.
View Full Product Details