Product Details
- SNP ID
-
rs1051130
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:41936044 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGCGCTGGGCTGGAGCTGGTCTGAG[A/C]GGCTTCCCTGAGGCTCTCCCTGAGT
- Phenotype
-
MIM: 603871
MIM: 123834
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BYSL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3218102] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BYSL
- Gene Name
- bystin like
There are no transcripts associated with this gene.
- Gene
- CCND3
- Gene Name
- cyclin D3
View Full Product Details