Product Details

SNP ID
rs1328865
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:8653454 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATATGGTCTTGGGAAGCTGAAAAT[A/G]CTAACTGAAACAAGCAGGACAGGGG
Phenotype
MIM: 612210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HULC PubMed Links

Gene Details

Gene
HULC
Gene Name
hepatocellular carcinoma up-regulated long non-coding RNA
There are no transcripts associated with this gene.

Gene
LOC100506207
Gene Name
uncharacterized LOC100506207
There are no transcripts associated with this gene.

View Full Product Details