Product Details

SNP ID
rs9512955
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:27987139 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCATCTGGCTGTCAAAGGCCATCCT[A/G]ATAGGCGGCATCGTCTGTTGGCTCC
Phenotype
MIM: 615804
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
URAD PubMed Links
Additional Information
For this assay, SNP(s) [rs73453643] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
URAD
Gene Name
ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5-) decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105577.1 Intron NP_001099047.1

View Full Product Details