Product Details

SNP ID
rs10759756
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:115147015 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGAGTCAGCTGGAGGTGCAGCAGC[A/G]CTTGAGCTATCCAAACTGCTAGGAT
Phenotype
MIM: 604767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEC1 PubMed Links

Gene Details

Gene
DEC1
Gene Name
deleted in esophageal cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017418.2 Intron NP_059114.1
Gene
LOC101928748
Gene Name
uncharacterized LOC101928748
There are no transcripts associated with this gene.

View Full Product Details