Product Details

SNP ID
rs10822055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:50993120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCGTGTCTGCCTCTGGAGTCCCTC[A/T]TGTGTATTGATTATATGGGATGGGG
Phenotype
MIM: 176894
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRKG1 PubMed Links

Gene Details

Gene
PRKG1
Gene Name
protein kinase, cGMP-dependent, type I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098512.2 Intron NP_001091982.1
NM_006258.3 Intron NP_006249.1
XM_011539952.2 Intron XP_011538254.1
XM_017016412.1 Intron XP_016871901.1
XM_017016413.1 Intron XP_016871902.1

View Full Product Details