Product Details
- SNP ID
-
rs2765499
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
44
- Location
-
Chr.1:157832924 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGGTGAAATAATTTTCAGAACGCC[A/T]CTTGTTTTTACATTCAATACCTGTA
- Phenotype
-
MIM: 602592
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CD5L
PubMed Links
Gene Details
- Gene
- CD5L
- Gene Name
- CD5 molecule like
View Full Product Details