Product Details

SNP ID
rs1757321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121569480 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTGATCAGCTCAGGCTGCTGTAA[C/T]GAAATAGACAAGGTGACTCACACAA
Phenotype
MIM: 609205
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAB2IP PubMed Links

Gene Details

Gene
DAB2IP
Gene Name
DAB2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032552.3 Intron NP_115941.2
NM_138709.2 Intron NP_619723.1
XM_005251719.4 Intron XP_005251776.1
XM_005251721.1 Intron XP_005251778.1
XM_005251724.4 Intron XP_005251781.1
XM_011518264.2 Intron XP_011516566.1
XM_011518265.2 Intron XP_011516567.1
XM_011518266.2 Intron XP_011516568.1
XM_011518267.2 Intron XP_011516569.1
XM_011518270.2 Intron XP_011516572.1
XM_011518271.2 Intron XP_011516573.1
XM_017014298.1 Intron XP_016869787.1
XM_017014299.1 Intron XP_016869788.1
XM_017014300.1 Intron XP_016869789.1

View Full Product Details