Product Details

SNP ID
rs6908950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42758611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGATTTACACTACAATGGAAATCT[C/G]GAGTGCTGAAATTATACAGAGTAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLTSCR1L PubMed Links

Gene Details

Gene
GLTSCR1L
Gene Name
GLTSCR1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318819.1 Intron NP_001305748.1
NM_015349.2 Intron NP_056164.1

View Full Product Details