Product Details

SNP ID
rs11191385
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:102753292 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGTGGCATACAGTAGACAGGCTGAG[G/T]TCTGGTCTGTGGGCACCTCCCTTTT
Phenotype
MIM: 611129
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WBP1L PubMed Links

Gene Details

Gene
WBP1L
Gene Name
WW domain binding protein 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083913.1 Intron NP_001077382.1
NM_017787.4 Intron NP_060257.4
XM_011539913.2 Intron XP_011538215.1
XM_017016360.1 Intron XP_016871849.1

View Full Product Details