Product Details
- SNP ID
-
rs7790688
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130380728 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCACCACCTGGGACAGCTGGCAGAT[A/G]AGCAACCAGGGTTGGGAAGAGGTTG
- Phenotype
-
MIM: 114850
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPA1
PubMed Links
Gene Details
- Gene
- CPA1
- Gene Name
- carboxypeptidase A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001868.3 |
|
Intron |
|
|
NP_001859.1 |
View Full Product Details