Product Details

SNP ID
rs1348310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4102115 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGGTTTTTATCTTGGGTTCCTT[C/G]TTTCATGTGTTTCTTTCCATTTGTC
Phenotype
MIM: 180410 MIM: 605921
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RRM1 PubMed Links

Gene Details

Gene
RRM1
Gene Name
ribonucleotide reductase catalytic subunit M1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033.4 Intron NP_001024.1
NM_001318064.1 Intron NP_001304993.1
NM_001318065.1 Intron NP_001304994.1
XM_011520278.1 Intron XP_011518580.1
Gene
STIM1
Gene Name
stromal interaction molecule 1
There are no transcripts associated with this gene.

View Full Product Details