Product Details

SNP ID
rs1401982
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:89595822 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTAACGAGATATGATTTACTCCCTG[G/A]ATAACTCCAAGAACATAGGACACAG
Phenotype
MIM: 108731
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ATP2B1 PubMed Links

Gene Details

Gene
ATP2B1
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001323.1 Intron NP_001001323.1
NM_001682.2 Intron NP_001673.2
XM_011538407.2 Intron XP_011536709.1
XM_017019357.1 Intron XP_016874846.1
XM_017019358.1 Intron XP_016874847.1
XM_017019359.1 Intron XP_016874848.1
XM_017019360.1 Intron XP_016874849.1

View Full Product Details