Product Details

SNP ID
rs12008134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48576728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTACCTAAAATGAGTATGAAAT[A/T]TAGCATTTTAAGTTAATTGTGTAGT
Phenotype
MIM: 300027
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RBM3 PubMed Links

Gene Details

Gene
RBM3
Gene Name
RNA binding motif (RNP1, RRM) protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006743.4 Intron NP_006734.1

View Full Product Details