Product Details
- SNP ID
-
rs2745357
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:127126444 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTAAGTAGTTTAATATATCATTTT[A/C]AACTGTGGTTTTTTATGGTTTTTAT
- Phenotype
-
MIM: 610574
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RSPO3
PubMed Links
Gene Details
- Gene
- RSPO3
- Gene Name
- R-spondin 3
View Full Product Details