Product Details
- SNP ID
-
rs2077947
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:135238353 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAGCCCTGGCGGCTCCTAGAGGCGA[C/G]TGGTGGGAAGTCTGCCCCCTATGGG
- Phenotype
-
MIM: 609987
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
STRA8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117917236] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- STRA8
- Gene Name
- stimulated by retinoic acid 8
View Full Product Details