Product Details

SNP ID
rs2236246
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:103912869 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATATGAAACACCCAGAGTGCTTAAG[G/T]GTTCATGTAATGACAGCAGACAGGC
Phenotype
MIM: 604573
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C14orf2 PubMed Links

Gene Details

Gene
C14orf2
Gene Name
chromosome 14 open reading frame 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127393.1 Intron NP_001120865.1
NM_004894.2 Intron NP_004885.1

View Full Product Details