Product Details

SNP ID
rs12492643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:105659029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCAAATTCTCGGAGGATGCTCC[A/G]GGCAACTTCGACATTATTCTGGGCT
Phenotype
MIM: 604491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLB PubMed Links

Gene Details

Gene
CBLB
Gene Name
Cbl proto-oncogene B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321786.1 2776 Missense Mutation NP_001308715.1
NM_001321788.1 2776 Missense Mutation NP_001308717.1
NM_001321789.1 2776 Missense Mutation NP_001308718.1
NM_001321790.1 2776 Missense Mutation NP_001308719.1
NM_001321791.1 2776 Missense Mutation NP_001308720.1
NM_001321793.1 2776 Missense Mutation NP_001308722.1
NM_001321794.1 2776 Missense Mutation NP_001308723.1
NM_001321795.1 2776 Missense Mutation NP_001308724.1
NM_001321796.1 2776 Missense Mutation NP_001308725.1
NM_001321797.1 2776 Missense Mutation NP_001308726.1
NM_001321798.1 2776 Missense Mutation NP_001308727.1
NM_001321799.1 2776 Missense Mutation NP_001308728.1
NM_001321806.1 2776 Missense Mutation NP_001308735.1
NM_001321807.1 2776 Missense Mutation NP_001308736.1
NM_001321808.1 2776 Missense Mutation NP_001308737.1
NM_001321811.1 2776 Missense Mutation NP_001308740.1
NM_001321813.1 2776 Missense Mutation NP_001308742.1
NM_001321816.1 2776 Missense Mutation NP_001308745.1
NM_001321820.1 2776 Missense Mutation NP_001308749.1
NM_001321822.1 2776 Missense Mutation NP_001308751.1
NM_170662.4 2776 Missense Mutation NP_733762.2
XM_011513257.1 2776 Missense Mutation XP_011511559.1
XM_011513259.2 2776 Intron XP_011511561.2
XM_017007395.1 2776 Missense Mutation XP_016862884.1
XM_017007396.1 2776 Missense Mutation XP_016862885.1
XM_017007397.1 2776 Missense Mutation XP_016862886.1
XM_017007398.1 2776 Missense Mutation XP_016862887.1
XM_017007399.1 2776 Missense Mutation XP_016862888.1
XM_017007400.1 2776 Missense Mutation XP_016862889.1

View Full Product Details