Product Details
- SNP ID
-
rs7250447
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57807755 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCATGTAGCAATCCTCCCACATGA[A/C]AGCTCTCCTGTGAGTTATGCAGCCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF552
PubMed Links
Gene Details
- Gene
- ZNF552
- Gene Name
- zinc finger protein 552
View Full Product Details