Product Details

SNP ID
rs11745088
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53483680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACTCCTAAAGGCAAGATGTAAA[C/G]AGCAGCCAGAACTGGAAGTCCAGTA
Phenotype
MIM: 136470
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
FST PubMed Links

Gene Details

Gene
FST
Gene Name
follistatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006350.3 622 Missense Mutation CAG,GAG Q152E NP_006341.1
NM_013409.2 622 Missense Mutation CAG,GAG Q152E NP_037541.1
XM_005248400.4 622 Missense Mutation CAG,GAG Q152E XP_005248457.1
XM_005248401.4 622 Missense Mutation CAG,GAG Q152E XP_005248458.1
XM_005248402.4 622 Missense Mutation CAG,GAG Q152E XP_005248459.1
XM_005248403.4 622 Missense Mutation CAG,GAG Q152E XP_005248460.1
XM_011543099.2 622 Missense Mutation CAG,GAG Q147E XP_011541401.1
XM_017008954.1 622 Missense Mutation CAG,GAG Q152E XP_016864443.1
XM_017008955.1 622 Missense Mutation CAG,GAG Q74E XP_016864444.1

View Full Product Details