Product Details

SNP ID
rs1043381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95885633 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAGTGTCTGAATTGATGGACCA[C/T]TTTTGTCTAAAACTAATTTCAACAT
Phenotype
MIM: 601874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELL2 PubMed Links

Gene Details

Gene
ELL2
Gene Name
elongation factor for RNA polymerase II 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012081.5 5778 UTR 3 NP_036213.2
XM_006714575.2 5778 UTR 3 XP_006714638.1
XM_017009239.1 5778 Intron XP_016864728.1
XM_017009240.1 5778 UTR 3 XP_016864729.1
XM_017009241.1 5778 UTR 3 XP_016864730.1
XM_017009242.1 5778 UTR 3 XP_016864731.1
XM_017009243.1 5778 UTR 3 XP_016864732.1

View Full Product Details