Product Details
- SNP ID
-
rs26682
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:52993936 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGCAAGTGAAAGAAATGGAAGGT[G/T]GGTGGAGTTCTTTTTTTCCAGTTGT
- Phenotype
-
MIM: 192974
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ITGA2
PubMed Links
Gene Details
- Gene
- ITGA2
- Gene Name
- integrin subunit alpha 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002203.3 |
|
Intron |
|
|
NP_002194.2 |
View Full Product Details