Product Details

SNP ID
rs2287980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70153367 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAATTCTGAAGGTCTGCTGCTTA[G/T]GAACTTTCTGTCTCTTCCTATAGTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC400541 PubMed Links

Gene Details

Gene
LOC400541
Gene Name
uncharacterized LOC400541
There are no transcripts associated with this gene.

Gene
PDPR
Gene Name
pyruvate dehydrogenase phosphatase regulatory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322117.1 Intron NP_001309046.1
NM_001322118.1 Intron NP_001309047.1
NM_001322119.1 Intron NP_001309048.1
NM_017990.4 Intron NP_060460.4
XM_005256015.2 Intron XP_005256072.1
XM_005256016.3 Intron XP_005256073.1
XM_006721224.2 Intron XP_006721287.1
XM_006721225.3 Intron XP_006721288.1
XM_011523190.2 Intron XP_011521492.1
XM_017023387.1 Intron XP_016878876.1
XM_017023388.1 Intron XP_016878877.1
XM_017023389.1 Intron XP_016878878.1

View Full Product Details