Product Details
- SNP ID
-
rs408028
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11768914 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TATCCCTAAGGACAGGAAAACCAAC[C/T]CCACTGAGGTGGTGCAAGAACCTGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF441
PubMed Links
Gene Details
- Gene
- ZNF441
- Gene Name
- zinc finger protein 441
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152355.2 |
|
Intron |
|
|
NP_689568.2 |
View Full Product Details