Product Details

SNP ID
rs36054492
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51331683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAGCCCAACTCTTCACTGGCAC[A/C]AAAACATTTGATCAGTCAACAAATA
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
IGLON5 PubMed Links

Gene Details

Gene
IGLON5
Gene Name
IgLON family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101372.1 3532 Intron NP_001094842.1
Gene
VSIG10L
Gene Name
V-set and immunoglobulin domain containing 10 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163922.1 3532 UTR 3 NP_001157394.1

View Full Product Details