Product Details

SNP ID
rs3094295
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:101683151 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAATTGAACTCTAAGGAAAATGG[C/T]TATTCCATGAACTGCATAGTAGTAT
Phenotype
MIM: 604180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105374013 PubMed Links

Gene Details

Gene
LOC105374013
Gene Name
endogenous retrovirus group K member 5 Gag polyprotein
There are no transcripts associated with this gene.

Gene
RPL24
Gene Name
ribosomal protein L24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000986.3 Intron NP_000977.1
Gene
ZBTB11
Gene Name
zinc finger and BTB domain containing 11
There are no transcripts associated with this gene.

Gene
ZBTB11-AS1
Gene Name
ZBTB11 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details