Product Details
- SNP ID
-
rs822395
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:186849018 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATCGCACCTATTAGTGGAGAAAT[A/C]TGTCCATAATACTGAAGTTTGGGGA
- Phenotype
-
MIM: 605441
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADIPOQ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115867072] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADIPOQ
- Gene Name
- adiponectin, C1Q and collagen domain containing
- Gene
- ADIPOQ-AS1
- Gene Name
- ADIPOQ antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details