Product Details

SNP ID
hCV2916296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54140254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGTGTCCTGCTCACCTCCCCCC[A/G]CCCCGCCACCCGCCTTGTGGGTCCC
Phenotype
MIM: 604910 MIM: 606419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT3 PubMed Links

Gene Details

Gene
CNOT3
Gene Name
CCR4-NOT transcription complex subunit 3
There are no transcripts associated with this gene.

Gene
PRPF31
Gene Name
pre-mRNA processing factor 31
There are no transcripts associated with this gene.

View Full Product Details