Product Details

SNP ID
rs12979251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17487021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGGCCCCTGGGCGGGCGGGGA[G/T]ATGCTGCGCCCCAGGCCTCGGAAGG
Phenotype
MIM: 600691
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC27A1 PubMed Links

Gene Details

Gene
SLC27A1
Gene Name
solute carrier family 27 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198580.2 Intron NP_940982.1
XM_011528000.1 Intron XP_011526302.1
XM_011528001.2 Intron XP_011526303.1
XM_011528002.2 Intron XP_011526304.1
XM_011528003.2 Intron XP_011526305.1
XM_017026781.1 Intron XP_016882270.1

View Full Product Details