Product Details

SNP ID
rs6826373
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:41744443 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACGGTCACGTAGAGGAGACAGTCTG[C/T]ACTGATATTAACACGATACAATTGA
Phenotype
MIM: 603851
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHOX2B PubMed Links

Gene Details

Gene
PHOX2B
Gene Name
paired like homeobox 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003924.3 2669 UTR 3 NP_003915.2

View Full Product Details