Product Details
- SNP ID
-
rs11079787
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:47735859 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCACTCTGTTGTGTGGTCAGGAGG[A/C]CTTCCCACCCTGGGCCCTTGCTGGG
- Phenotype
-
MIM: 604895
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBX21
PubMed Links
Gene Details
- Gene
- TBX21
- Gene Name
- T-box 21
View Full Product Details