Product Details

SNP ID
rs34790642
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:160351579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTGCTGTCTTTGGTTCAACTA[A/G]TTTTTTTCTTTCAAATTCTGTCTGA
Phenotype
MIM: 604842
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SLC22A3 PubMed Links

Gene Details

Gene
SLC22A3
Gene Name
solute carrier family 22 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021977.3 Intron NP_068812.1
XM_005267106.4 Intron XP_005267163.1
XM_005267107.3 Intron XP_005267164.1
XM_011536075.2 Intron XP_011534377.1
XM_011536076.2 Intron XP_011534378.1
XM_017011203.1 Intron XP_016866692.1

View Full Product Details