Product Details

SNP ID
rs11796780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103505159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTTTTGCTTAATATTATCCTTG[C/T]GAGATATATCCATGATTTTGCATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB40A PubMed Links

Gene Details

Gene
RAB40A
Gene Name
RAB40A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080879.2 Intron NP_543155.2
XM_005262083.4 Intron XP_005262140.1
XM_011530872.2 Intron XP_011529174.1

View Full Product Details