Product Details

SNP ID
rs10769748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6997347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCACATTTTCATCTGTTATAGCAC[A/G]TAATTAGAAATTTTTAACCCAGTGT
Phenotype
MIM: 605015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF214 PubMed Links

Gene Details

Gene
ZNF214
Gene Name
zinc finger protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013249.2 4714 Intron NP_037381.2
XM_005253128.3 4714 UTR 3 XP_005253185.1
XM_005253129.3 4714 UTR 3 XP_005253186.1
XM_006718308.3 4714 UTR 3 XP_006718371.1
XM_006718309.3 4714 UTR 3 XP_006718372.1
XM_011520355.2 4714 UTR 3 XP_011518657.1
XM_011520357.2 4714 UTR 3 XP_011518659.1
XM_017018269.1 4714 UTR 3 XP_016873758.1

View Full Product Details