Product Details

SNP ID
rs1255396
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:42543739 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGGAATAGGTAATTCCAAAATTC[T/C]ATCCCAAACTAAAGCAACAATTAAG
Phenotype
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
ZNF37BP PubMed Links
Additional Information
For this assay, SNP(s) [rs73262610] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ZNF37BP
Gene Name
zinc finger protein 37B, pseudogene
There are no transcripts associated with this gene.

View Full Product Details