Product Details

SNP ID
rs11909619
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43672045 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTATTTCATCTCTAGTTCATCTG[A/C]ACTGATAATTAGAGATGCTTTTTTA
Phenotype
MIM: 610654
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
RRP1B PubMed Links

Gene Details

Gene
RRP1B
Gene Name
ribosomal RNA processing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015056.2 Intron NP_055871.1

View Full Product Details