Product Details

SNP ID
rs969060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43660971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGAGGTTTTAAGATAAAGTTT[G/T]ATTTAGTTACTGATGGTACTAATAC
Phenotype
MIM: 604554 MIM: 610654
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSF2BP PubMed Links

Gene Details

Gene
HSF2BP
Gene Name
heat shock transcription factor 2 binding protein
There are no transcripts associated with this gene.

Gene
RRP1B
Gene Name
ribosomal RNA processing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015056.2 Intron NP_055871.1

View Full Product Details