Product Details

SNP ID
rs396801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132725980 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGCCTGGCCTGGAAGCAGGGGA[C/G]AAAGGGACCCTCTACTCTACTGTGG
Phenotype
MIM: 615365
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AK8 PubMed Links

Gene Details

Gene
AK8
Gene Name
adenylate kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317958.1 Intron NP_001304887.1
NM_001317959.1 Intron NP_001304888.1
NM_152572.2 Intron NP_689785.1
XM_005272169.2 Intron XP_005272226.1
XM_006716965.2 Intron XP_006717028.1
XM_011518277.2 Intron XP_011516579.1
XM_011518278.2 Intron XP_011516580.1
XM_017014308.1 Intron XP_016869797.1

View Full Product Details