Product Details

SNP ID
rs3761822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133210436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCTCCCACGGCAAGTGCTGCC[A/G]AGCAGAAACCCCCCACCCTCATCCC
Phenotype
MIM: 604606
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OBP2B PubMed Links

Gene Details

Gene
OBP2B
Gene Name
odorant binding protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288987.1 Intron NP_001275916.1
NM_014581.3 Intron NP_055396.1
XM_006717086.3 Intron XP_006717149.1
XM_017014654.1 Intron XP_016870143.1
XM_017014655.1 Intron XP_016870144.1

View Full Product Details