Product Details

SNP ID
rs667913
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:120476229 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACTAAGCAGATTTTTAATGAACTG[C/T]GTCAATATAATTCAGTAATGGCACC
Phenotype
MIM: 601562 MIM: 601943
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNLL1 PubMed Links

Gene Details

Gene
DYNLL1
Gene Name
dynein light chain LC8-type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037494.1 Intron NP_001032583.1
NM_001037495.1 Intron NP_001032584.1
NM_003746.2 Intron NP_003737.1
Gene
SRSF9
Gene Name
serine and arginine rich splicing factor 9
There are no transcripts associated with this gene.

View Full Product Details