Product Details
- SNP ID
-
rs2245947
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:43132245 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATACTTAATATTGGCGCAGGAGCT[G/T]CTCTGAGGACGGGGCATGGAGCCAG
- Phenotype
-
MIM: 603392
MIM: 614479
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BIK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77274850] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BIK
- Gene Name
- BCL2 interacting killer
There are no transcripts associated with this gene.
- Gene
- MCAT
- Gene Name
- malonyl-CoA-acyl carrier protein transacylase
View Full Product Details