Product Details

SNP ID
rs485288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69586248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGTATTGGTACTTAAACGGAAT[A/G]CCTCTTGGTTTTCCTTTTCAGGCGT
Phenotype
MIM: 605139 MIM: 607743
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT2 PubMed Links

Gene Details

Gene
CCT2
Gene Name
chaperonin containing TCP1 subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198842.1 Intron NP_001185771.1
NM_006431.2 Intron NP_006422.1
Gene
FRS2
Gene Name
fibroblast growth factor receptor substrate 2
There are no transcripts associated with this gene.

Gene
MIR3913-1
Gene Name
microRNA 3913-1
There are no transcripts associated with this gene.

Gene
MIR3913-2
Gene Name
microRNA 3913-2
There are no transcripts associated with this gene.

View Full Product Details