Product Details
- SNP ID
-
rs9489437
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:118553182 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTTGTATGCTTTCCATCGAAAAA[G/T]TACTCTATTAAACAGCTTAGCTGTT
- Phenotype
-
MIM: 172405
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP85L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113254931] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP85L
- Gene Name
- centrosomal protein 85 like
- Gene
- PLN
- Gene Name
- phospholamban
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002667.4 |
|
Intron |
|
|
NP_002658.1 |
View Full Product Details