Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001207010.1 | Intron | NP_001193939.1 | |||
NM_001300730.1 | Intron | NP_001287659.1 | |||
NM_138337.5 | Intron | NP_612210.4 | |||
NM_201623.3 | Intron | NP_963917.2 | |||
XM_005253324.2 | Intron | XP_005253381.1 | |||
XM_006719033.3 | Intron | XP_006719096.1 | |||
XM_006719035.3 | Intron | XP_006719098.1 | |||
XM_006719036.3 | Intron | XP_006719099.1 | |||
XM_011520570.1 | Intron | XP_011518872.1 | |||
XM_011520571.2 | Intron | XP_011518873.2 | |||
XM_011520573.2 | Intron | XP_011518875.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001099431.1 | Intron | NP_001092901.1 | |||
NM_016509.3 | Intron | NP_057593.3 | |||
XM_005253382.4 | Intron | XP_005253439.1 | |||
XM_011520685.2 | Intron | XP_011518987.1 | |||
XM_011520686.2 | Intron | XP_011518988.1 | |||
XM_017019395.1 | Intron | XP_016874884.1 | |||
XM_017019396.1 | Intron | XP_016874885.1 |