Product Details

SNP ID
rs1456169
Assay Type
Validated
NCBI dbSNP Submissions
69
Location
Chr.1:99268408 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCAGCCCATTTGCAGAGTGTCAA[G/T]GTATCTCTAATTATACCCCAATGGG
Phenotype
MIM: 607813
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLPPR4 PubMed Links

Gene Details

Gene
PLPPR4
Gene Name
phospholipid phosphatase related 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166252.1 Intron NP_001159724.1
NM_014839.4 Intron NP_055654.2
XM_011542498.2 Intron XP_011540800.1

View Full Product Details