Product Details

SNP ID
rs11749024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:7862553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTTAAGTAAGTCTTTGAGGTCA[A/G]TATTACTATCATTTTACAGAGGAAG
Phenotype
MIM: 602568
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FASTKD3 PubMed Links

Gene Details

Gene
FASTKD3
Gene Name
FAST kinase domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024091.3 Intron NP_076996.2
XM_006714498.1 Intron XP_006714561.1
Gene
MTRR
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002454.2 Intron NP_002445.2
NM_024010.2 Intron NP_076915.2

View Full Product Details